4-(Hydroxymethyl)-1H-benzimidazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1H-Benzimidazol-4-yl)methanol, 4-(Hydroxymethyl)benzimidazole
A175558-100MG
A175558-1G
A175558-250MG
77029-234EA
22
USD
77029-234
77029-236
77029-238
4-(Hydroxymethyl)-1H-benzimidazole 97%
4-(Hydroxymethyl)-1H-benzimidazole
Precaución: for Research Use Only
Formula:
C₈H₈N₂O |
MDL Number:
MFCD15144561 CAS Number: 65658-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...