Fmoc-phenylalaninol 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phenylalaninol, N-Fmoc-L-phenylalaninol

A175523-1G A175523-250MG A175523-25G A175523-5G
76998-346EA 13.8 USD
76998-346 76998-348 76998-350 76998-352
Fmoc-phenylalaninol 98%
Fmoc-phenylalaninol

Precaución: for Research Use Only

Formula: C₂₄H₂₃NO₃
MW: 373.452 g/mol
MDL Number: MFCD00235941
CAS Number: 129397-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR