Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I) 98%
Supplier: Ambeed
|
Warning
|
A175422-100MG
A175422-1G
A175422-250MG
A175422-5G
76985-612EA
54
USD
76985-612
76985-614
76985-616
76985-618
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I) 98%
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I)
Precaución: for Research Use Only
Formula:
C₂₇H₃₆AuClN₂ |
MDL Number:
MFCD09839143 CAS Number: 852445-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...