Z-Thr(tBu)-OH·DCHA 97%

Supplier: Ambeed
Warning

A175081-100G A175081-5G
76998-342EA 229 USD
76998-342 77881-337
Z-Thr(tBu)-OH·DCHA 97%
Z-Thr(tBu)-OH·DCHA

Precaución: for Research Use Only

Formula: C₂₈H₄₆N₂O₅
MDL Number: MFCD00077043
CAS Number: 16966-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR