Methyl 3-chloro-6-oxo-1,6-dihydropyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A175004-100MG A175004-1G A175004-250MG
77011-726EA 30.77 USD
77011-726 77011-728 77011-730
Methyl 3-chloro-6-oxo-1,6-dihydropyridine-2-carboxylate 95%
Methyl 3-chloro-6-oxo-1,6-dihydropyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
CAS Number: 1214361-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR