4-Iododibenzothiophene 98%

Supplier: Ambeed
Warning

Synonyms: 4-Iododibenzo[b,d]thiophene

A174464-250MG A174464-25G A174464-5G A174464-1G
76985-588EA 9.55 USD
76985-588 76985-590 76985-592 77611-698
4-Iododibenzothiophene 98%
4-Iododibenzothiophene

Precaución: for Research Use Only

Formula: C₁₂H₇IS
MW: 310.15 g/mol
MDL Number: MFCD00159895
CAS Number: 132034-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR