DL-3-Pyrrolidinol 97%

Supplier: Ambeed
Warning

Synonyms: Pyrrolidin-3-ol, 3-Pyrrolidinol, DL-3-Hydroxypyrrolidine

A174388-100G A174388-1G A174388-5G A174388-10G A174388-25G
77029-220EA 313 USD
77029-220 77029-222 77029-224 77671-472 77871-871
DL-3-Pyrrolidinol 97%
DL-3-Pyrrolidinol

Precaución: for Research Use Only

Formula: C₄H₉NO
MDL Number: MFCD00005256
CAS Number: 40499-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR