N-Boc-3-Amino-4-iodopyridine 96%

Supplier: Ambeed
Warning

A174361-100MG A174361-1G A174361-250MG A174361-5G
77011-704EA 18 USD
77011-704 77011-706 77011-708 77011-710
N-Boc-3-Amino-4-iodopyridine 96%
N-Boc-3-Amino-4-iodopyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃IN₂O₂
MDL Number: MFCD04973410
CAS Number: 154048-89-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR