1-(2-Amino-3-chlorophenyl)ethanone 95%

Supplier: Ambeed
Warning

A174349-100MG A174349-1G A174349-250MG
76985-582EA 29.71 USD
76985-582 76985-584 76985-586
1-(2-Amino-3-chlorophenyl)ethanone 95%
1-(2-Amino-3-chlorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MW: 169.60 g/mol
MDL Number: MFCD11113372
CAS Number: 56762-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR