Oxybenzone 98%

Supplier: Ambeed
Warning

Synonyms: Eusolex® 4360, 2-Hydroxy-4-methoxybenzophenone, (2-Hydroxy-4-methoxyphenyl)(phenyl)methanone

A174222-25G A174222-500G A174222-100G
77029-216EA 6.37 USD
77029-216 77029-218 77600-414
Oxybenzone 98%
Oxybenzone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD00008387
CAS Number: 131-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR