2-Isopropyl-4-methylanisole 95%

Supplier: Ambeed
Warning

A174214-1G A174214-250MG
76985-572EA 150 USD
76985-572 76985-574
2-Isopropyl-4-methylanisole 95%
2-Isopropyl-4-methylanisole

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
MDL Number: MFCD18967937
CAS Number: 31574-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR