2-Chloro-4-trifluoromethoxy-benzaldehyde 96%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-(trifluoromethoxy)benzaldehyde

A173739-100MG A173739-1G A173739-250MG
77011-690EA 61.54 USD
77011-690 77011-692 77011-694
2-Chloro-4-trifluoromethoxy-benzaldehyde 96%
2-Chloro-4-trifluoromethoxy-benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.564 g/mol
MDL Number: MFCD18393768
CAS Number: 1079351-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR