1-Phenyl-2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-ol 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl-PEG9-alcohol

A173556-1G A173556-250MG A173556-100MG A173556-5G
77011-686EA 168 USD
77011-686 77011-688 77890-117 77890-118
1-Phenyl-2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-ol 98%
1-Phenyl-2,5,8,11,14,17,20,23,26-nonaoxaoctacosan-28-ol

Precaución: for Research Use Only

Formula: C₂₅H₄₄O₁₀
MDL Number: MFCD24682797
CAS Number: 868594-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR