1-Phenylpiperidine-3-carboxamide 95%

Supplier: Ambeed
Warning

A173535-100MG A173535-1G A173535-250MG
77011-680EA 27.59 USD
77011-680 77011-682 77011-684
1-Phenylpiperidine-3-carboxamide 95%
1-Phenylpiperidine-3-carboxamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O
MW: 204.27 g/mol
MDL Number: MFCD24628624
CAS Number: 58971-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR