2-(2-Hydroxyethyl)pyridine 95%

Supplier: Ambeed
Warning

A173502-100G A173502-25G A173502-500G A173502-10G A173502-1000G
77029-196EA 40 USD
77029-196 77029-198 77029-200 77593-088 77671-466
2-(2-Hydroxyethyl)pyridine 95%
2-(2-Hydroxyethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD00006364
CAS Number: 103-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR