8-Bromo-7-quinolinol 97%

Supplier: Ambeed
Warning

A173417-100MG A173417-250MG A173417-1G A173417-5G A173417-10G
77029-188EA 11 USD
77029-188 77029-190 77600-406 77667-414 77871-865
8-Bromo-7-quinolinol 97%
8-Bromo-7-quinolinol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MDL Number: MFCD13188513
CAS Number: 90224-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR