3-(2-Aminoethyl)oxazolidin-2-one hydrochloride 95%

Supplier: Ambeed
Warning

A173333-100MG A173333-250MG
77011-676EA 116.7 USD
77011-676 77011-678
3-(2-Aminoethyl)oxazolidin-2-one hydrochloride 95%
3-(2-Aminoethyl)oxazolidin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClN₂O₂
MW: 166.60 g/mol
MDL Number: MFCD13193842
CAS Number: 1262773-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR