N-(tert-Butyl)hydroxylamine acetate 95%

Supplier: Ambeed
Warning

A173321-1G A173321-250MG A173321-5G
76985-542EA 21.22 USD
76985-542 76985-544 76985-546
N-(tert-Butyl)hydroxylamine acetate 95%
N-(tert-Butyl)hydroxylamine acetate

Precaución: for Research Use Only

Formula: C₆H₁₅NO₃
MW: 149.18 g/mol
MDL Number: MFCD01321354
CAS Number: 253605-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR