3,5-Dichloro-2-methoxyaniline 95%

Supplier: Ambeed
Warning

A173210-100MG A173210-5G A173210-1G
77011-672EA 30 USD
77011-672 77011-674 77611-688
3,5-Dichloro-2-methoxyaniline 95%
3,5-Dichloro-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MDL Number: MFCD11977376
CAS Number: 33353-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR