Methyl-2-amino-4-fluorobenzoate 97%
Supplier: Ambeed
|
Warning
|
A173129-10G
A173129-1G
A173129-25G
A173129-5G
77029-174EA
53
USD
77029-174
77029-176
77029-178
77029-180
Methyl-2-amino-4-fluorobenzoate 97%
Methyl 2-amino-4-fluorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈FNO₂ |
MDL Number:
MFCD11054082 CAS Number: 2475-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...