3-Bromo-4-morpholinobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A173085-10G
A173085-1G
A173085-250MG
A173085-5G
77029-162EA
234.46
USD
77029-162
77029-164
77029-166
77029-168
3-Bromo-4-morpholinobenzoic acid 97%
3-Bromo-4-morpholinobenzoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₂BrNO₃ MW: 286.12 g/mol |
MDL Number:
MFCD11110891 CAS Number: 1131594-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...