(1S,2S)-2-((tert-Butoxycarbonyl)amino)cyclopropanecarboxylic acid 95%

Supplier: Ambeed
Warning

A173008-100MG A173008-250MG
76985-538EA 575.01 USD
76985-538 76985-540
(1S,2S)-2-((tert-Butoxycarbonyl)amino)cyclopropanecarboxylic acid 95%
(1S,2S)-2-((tert-Butoxycarbonyl)amino)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MW: 201.21 g/mol
MDL Number: MFCD09863454
CAS Number: 1486470-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR