MMAE 98%

Supplier: Ambeed
Danger

Synonyms: Monomethyl auristatin E, (S)-N-((3R,4S,5S)-1-((S)-2-((1R,2R)-3-(((1S,2R)-1-Hydroxy-1-phenylpropan-2-yl)amino)-1-methoxy-2-methyl-3-oxopropyl)pyrrolidin-1-yl)-3-methoxy-5-methyl-1-oxoheptan-4-yl)-N,3-dimethyl-2-((S)-3-methyl-2-(methylamino)butanamido)butanamide, N-Methyl-L-valyl-N-[(1S,2R)-4-[(2S)-2-[(1R,2R)-3-[[(1R,2S)-2-hydroxy-1-methyl-2-phenylethyl]amino]-1-methoxy-2-methyl-3-oxopropyl]-1-pyrrolidinyl]-2-methoxy-1-[(1S)-1-methylpropyl]-4-oxobutyl]-N-methyl-L-valinamide

A172961-1MG A172961-100MG A172961-1G A172961-250MG
76985-532EA 80.63 USD
76985-532 76985-534 76985-536 77567-548 77567-550 77567-552
MMAE 98%
MMAE

Precaución: For research use only.

Formula: C₃₉H₆₇N₅O₇
MDL Number: MFCD22124498
CAS Number: 474645-27-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR