Dihydro-2H-pyran-3(4H)-one 95%

Supplier: Ambeed
Warning

A172825-10G A172825-1G A172825-250MG A172825-5G
77011-650EA 103.97 USD
77011-650 77011-652 77011-654 77011-656
Dihydro-2H-pyran-3(4H)-one 95%
Dihydro-2H-pyran-3(4H)-one

Precaución: for Research Use Only

Formula: C₅H₈O₂
MW: 100.1158 g/mol
MDL Number: MFCD00182426
CAS Number: 23462-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR