N,N-Dimethyl-4-vinylaniline 97%

Supplier: Ambeed
Danger

A172819-100MG A172819-10G A172819-1G A172819-250MG A172819-25G A172819-5G
77029-140EA 12 USD
77029-140 77029-142 77029-144 77029-146 77029-148 77029-150
N,N-Dimethyl-4-vinylaniline 97%
N,N-Dimethyl-4-vinylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MW: 147.217 g/mol
MDL Number: MFCD00085252
CAS Number: 2039-80-7
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR