Ethyl 2-(3-formyl-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A172789-100G
A172789-100MG
A172789-10G
A172789-1G
A172789-250MG
A172789-25G
A172789-5G
77029-126EA
1590.29
USD
77029-126
77029-128
77029-130
77029-132
77029-134
77029-136
77029-138
Ethyl 2-(3-formyl-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylate 98%
Ethyl 2-(3-formyl-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₁₈H₂₁NO₄S MW: 347.43 g/mol |
MDL Number:
MFCD13194811 CAS Number: 161798-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...