5-Chloro-3-iodo-1H-pyrazolo[4,3-b]pyridine 95%

Supplier: Ambeed
Warning

A172747-100MG A172747-1G A172747-250MG A172747-5G
76985-520EA 21 USD
76985-520 76985-522 76985-524 76985-526
5-Chloro-3-iodo-1H-pyrazolo[4,3-b]pyridine 95%
5-Chloro-3-iodo-1H-pyrazolo[4,3-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClIN₃
MW: 279.46 g/mol
MDL Number: MFCD16658104
CAS Number: 1357945-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR