N-Methyl-N-((3R,4R)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride 97%

Supplier: Ambeed
Warning

A172604-100MG A172604-1G A172604-250MG A172604-5G
77029-118EA 22 USD
77029-118 77029-120 77029-122 77029-124
N-Methyl-N-((3R,4R)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride 97%
N-Methyl-N-((3R,4R)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₀ClN₅
MW: 281.78 g/mol
MDL Number: MFCD14706832
CAS Number: 1260590-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR