5-Amino-4,6-dichloro-2-methylpyrimidine 95%
Supplier: Ambeed
|
Warning
|
A172565-100G
A172565-10G
A172565-1G
A172565-25G
A172565-5G
77011-634EA
92
USD
77011-634
77011-636
77011-638
77011-640
77011-642
5-Amino-4,6-dichloro-2-methylpyrimidine 95%
5-Amino-4,6-dichloro-2-methylpyrimidine
Precaución: for Research Use Only
Formula:
C₅H₅Cl₂N₃ |
MDL Number:
MFCD00194053 CAS Number: 39906-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...