4,5-Dibromo-2-methyl-2H-1,2,3-triazole 97%

Supplier: Ambeed
Warning

A172490-100MG A172490-1G A172490-250MG A172490-5G A172490-100G A172490-25G
76985-506EA 5 USD
76985-506 76985-508 76985-510 76985-512 77890-111 77890-112
4,5-Dibromo-2-methyl-2H-1,2,3-triazole 97%
4,5-Dibromo-2-methyl-2H-1,2,3-triazole

Precaución: for Research Use Only

Formula: C₃H₃Br₂N₃
MDL Number: MFCD26383581
CAS Number: 28938-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR