4-Formyl-3-methylbenzonitrile 97%

Supplier: Ambeed
Warning

A172446-100MG A172446-1G
77011-630EA 20.16 USD
77011-630 77011-632
4-Formyl-3-methylbenzonitrile 97%
4-Formyl-3-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.15 g/mol
MDL Number: MFCD18394297
CAS Number: 27609-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR