L-Tryptophan ethyl ester hydrochloride 97%

Supplier: Ambeed
Warning

A172341-100G A172341-25G A172341-5G
76998-264EA 55 USD
76998-264 76998-266 76998-268 76998-270
L-Tryptophan ethyl ester hydrochloride 97%
L-Tryptophan ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂
MDL Number: MFCD00038993
CAS Number: 2899-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR