5,6-Dichloropyridazin-4-amine 97%
Supplier: Ambeed
|
Warning
|
A172338-100MG
A172338-250MG
A172338-1G
77029-110EA
41.38
USD
77029-110
77029-112
77567-534
5,6-Dichloropyridazin-4-amine 97%
5,6-Dichloropyridazin-4-amine
Precaución: For research use only.
Formula:
C₄H₃Cl₂N₃ MW: 163.99 g/mol |
MDL Number:
MFCD01647252 CAS Number: 89180-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...