tert-Butyl-2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 6,7-dihydro-2H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate, 2,4,6,7-Tetrahydro-5H-pyrazolo[4,3-c]pyridine, N5-BOC protected

A172283-10G A172283-1G A172283-250MG A172283-25G A172283-5G
76998-252EA 245.07 USD
76998-252 76998-254 76998-256 76998-258 76998-260
tert-Butyl-2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate 97%
tert-Butyl-2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃O₂
MW: 223.28 g/mol
MDL Number: MFCD08689720
CAS Number: 924869-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR