Ethyl 2-amino-4,5-bis(2-methoxyethoxy)benzoate 97%

Supplier: Ambeed
Warning

A172243-10G A172243-1G A172243-250MG A172243-5G
77029-096EA 65 USD
77029-096 77029-098 77029-100 77029-102
Ethyl 2-amino-4,5-bis(2-methoxyethoxy)benzoate 97%
Ethyl 2-amino-4,5-bis(2-methoxyethoxy)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₂₃NO₆
MDL Number: MFCD11618122
CAS Number: 179688-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR