9-(Bromomethyl)nonadecane 97%

Supplier: Ambeed
Warning

A172229-100G A172229-25G A172229-500G
76985-490EA 157 USD
76985-490 76985-492 76985-494
9-(Bromomethyl)nonadecane 97%
9-(Bromomethyl)nonadecane

Precaución: for Research Use Only

Formula: C₂₀H₄₁Br
MDL Number: MFCD28515435
CAS Number: 69620-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR