Pyrazine-2,5-diamine dihydrochloride 97%

Supplier: Ambeed
Warning

A172206-100MG A172206-1G A172206-250MG
76985-484EA 54.11 USD
76985-484 76985-486 76985-488
Pyrazine-2,5-diamine dihydrochloride 97%
Pyrazine-2,5-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₈Cl₂N₄
MW: 183.04 g/mol
MDL Number: MFCD26406397
CAS Number: 1588441-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR