(2-Amino-4,5-dimethylthiophen-3-yl)(4-chlorophenyl)methanone 98%

Supplier: Ambeed
Warning

A172152-1G A172152-250MG A172152-5G
77011-620EA 27.59 USD
77011-620 77011-622 77611-676
(2-Amino-4,5-dimethylthiophen-3-yl)(4-chlorophenyl)methanone 98%
(2-Amino-4,5-dimethylthiophen-3-yl)(4-chlorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNOS
MW: 265.76 g/mol
MDL Number: MFCD00177532
CAS Number: 50508-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR