(S)-2-(4-Methylphenylsulfonamido)propanoic acid 97%

Supplier: Ambeed
Warning

A172151-100G A172151-10G A172151-1G A172151-250MG A172151-25G A172151-5G
76985-472EA 130 USD
76985-472 76985-474 76985-476 76985-478 76985-480 76985-482
(S)-2-(4-Methylphenylsulfonamido)propanoic acid 97%
(S)-2-(4-Methylphenylsulfonamido)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄S
MDL Number: MFCD02259513
CAS Number: 21957-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR