4,5-Dimethoxy-1-benzocyclobutenecarbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile, 4,5-Dimethoxy-1-cyanobenzocyclobutane, 1-Cyano-4,5-dimethoxybenzocyclobutene

A172126-100G A172126-10G A172126-1G A172126-250MG A172126-25G A172126-5G
77029-084EA 359.65 USD
77029-084 77029-086 77029-088 77029-090 77029-092 77029-094
4,5-Dimethoxy-1-benzocyclobutenecarbonitrile 97%
4,5-Dimethoxy-1-benzocyclobutenecarbonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD01846132
CAS Number: 35202-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR