(S)-Methyl 2-((tert-butoxycarbonyl)amino)pent-4-ynoate 95%

Supplier: Ambeed
Warning

A172078-100MG A172078-1G A172078-250MG
76985-464EA 42.44 USD
76985-464 76985-466 76985-468
(S)-Methyl 2-((tert-butoxycarbonyl)amino)pent-4-ynoate 95%
(S)-Methyl 2-((tert-butoxycarbonyl)amino)pent-4-ynoate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MW: 227.25 g/mol
MDL Number: MFCD07368399
CAS Number: 71460-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR