1-(1-Methyl-1H-imidazol-2-yl)ethanone 95%

Supplier: Ambeed
Warning

A171979-100MG A171979-1G A171979-250MG A171979-5G
76998-244EA 16 USD
76998-244 76998-246 76998-248 76998-250
1-(1-Methyl-1H-imidazol-2-yl)ethanone 95%
1-(1-Methyl-1H-imidazol-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD08668178
CAS Number: 85692-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR