7-Dimethoxymethyl-1,2,3,4-tetrahydro[1,8]naphthyridine 97%

Supplier: Ambeed
Warning

Synonyms: 7-(Dimethoxymethyl)-1,2,3,4-tetrahydro-1,8-naphthyridine

A171947-100MG A171947-1G A171947-250MG A171947-5G
77011-612EA 41.38 USD
77011-612 77011-614 77011-616 77011-618
7-Dimethoxymethyl-1,2,3,4-tetrahydro[1,8]naphthyridine 97%
7-Dimethoxymethyl-1,2,3,4-tetrahydro[1,8]naphthyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₂
MW: 208.26 g/mol
MDL Number: MFCD06657573
CAS Number: 204452-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR