3,4-Dibromoanisole 95%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dibromo-4-methoxybenzene

A171886-100G A171886-10G A171886-1G A171886-25G A171886-5G
77029-068EA 218.55 USD
77029-068 77029-070 77029-072 77029-074 77029-076
3,4-Dibromoanisole 95%
3,4-Dibromoanisole

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
MW: 265.93 g/mol
MDL Number: MFCD00070786
CAS Number: 62415-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR