3-Bromophenyl acetate 98%

Supplier: Ambeed
Warning

A171846-1G A171846-5G
77029-064EA 21.22 USD
77029-064 77029-066
3-Bromophenyl acetate 98%
3-Bromophenyl acetate

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MW: 215.04 g/mol
MDL Number: MFCD08063242
CAS Number: 35065-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR