2-Ethoxy-5-methoxybenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxy-5-methoxyphenylboronic acid

A171836-1G A171836-250MG A171836-5G
77029-058EA 18 USD
77029-058 77029-060 77029-062
2-Ethoxy-5-methoxybenzeneboronic acid 97%
2-Ethoxy-5-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₄
MDL Number: MFCD09878346
CAS Number: 957065-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR