3-Ethynylbenzoic acid 97%

Supplier: Ambeed
Warning

A171764-1G A171764-250MG A171764-25G A171764-5G
77011-604EA 19 USD
77011-604 77011-606 77011-608 77011-610
3-Ethynylbenzoic acid 97%
3-Ethynylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₆O₂
MDL Number: MFCD06658448
CAS Number: 10601-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR