6-Nitro-3H-imidazo[4,5-b]pyridine 95%

Supplier: Ambeed
Warning

A171760-100MG A171760-250MG A171760-1G
77029-052EA 58 USD
77029-052 77029-054 77667-396
6-Nitro-3H-imidazo[4,5-b]pyridine 95%
6-Nitro-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄N₄O₂
MDL Number: MFCD11215559
CAS Number: 3537-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR