2'-Deoxy-2'-fluoroarabinofuranosylcytosine, [3H] 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1-((2R,3S,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

A171736-100MG A171736-1G A171736-250MG A171736-25MG
76985-456EA 123.07 USD
76985-456 76985-458 76985-460 76985-462
2'-Deoxy-2'-fluoroarabinofuranosylcytosine, [3H] 95%
2'-Deoxy-2'-fluoroarabinofuranosylcytosine, [3H]

Precaución: for Research Use Only

Formula: C₉H₁₂FN₃O₄
MW: 245.21 g/mol
MDL Number: MFCD07374399
CAS Number: 56632-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR