(R)-2-((tert-Butoxycarbonyl)amino)-5-methylhexanoic acid 97%
Supplier: Ambeed
|
Warning
|
A171560-100MG
A171560-1G
A171560-250MG
77011-594EA
40.32
USD
77011-594
77011-596
77011-598
(R)-2-((tert-Butoxycarbonyl)amino)-5-methylhexanoic acid 97%
(R)-2-((tert-Butoxycarbonyl)amino)-5-methylhexanoic acid
Precaución: for Research Use Only
Formula:
C₁₂H₂₃NO₄ MW: 245.32 g/mol |
MDL Number:
MFCD11973692 CAS Number: 1445976-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...